Learn More
Medchemexpress LLC Retinoic acid-d5 | 78996-15-3 | 99.1% | 305.47 | 500 UG
SDP
Catalog No. 5000362573
Encompass_Preferred
Retinoic acid-d5 is a deuterium-labeled Retinoic acid, a metabolite of vitamin A crucial for cell growth, differentiation, and organogenesis. It acts as a natural agonist of RAR nuclear receptors with IC50s of 14 nM for RARα/β/γ, and binds to PPARβ/δ with a Kd of 17 nM. It also inhibits transcription factor Nrf2 through retinoic acid receptor alpha activation.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62573 Supplier Medchemexpress LLC Supplier No. HY14649S4500UG
Retinoic acid-d5 is a deuterium-labeled Retinoic acid, a metabolite of vitamin A crucial for cell growth, differentiation, and organogenesis. It acts as a natural agonist of RAR nuclear receptors with IC50s of 14 nM for RARα/β/γ, and binds to PPARβ/δ with a Kd of 17 nM. It also inhibits transcription factor Nrf2 through retinoic acid receptor alpha activation.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS