Learn More
Medchemexpress LLC Retinoic acid-d6 | 2483831-72-5 | 98.9% | 306.47 | 1 MG
SDP
Catalog No. 5000360753
Encompass_Preferred
Retinoic acid-d6 is the deuterium labeled Retinoic acid, a metabolite of vitamin A crucial for cell growth, differentiation, and organogenesis. It acts as a natural agonist of RAR nuclear receptors (IC50s of 14 nM for RARα/β/γ) and binds to PPARβ/δ with a Kd of 17 nM. This compound inhibits transcription factor Nrf2 through activation of retinoic acid receptor alpha.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60753 Supplier Medchemexpress LLC Supplier No. HY14649S31MG
Retinoic acid-d6 is the deuterium labeled Retinoic acid, a metabolite of vitamin A crucial for cell growth, differentiation, and organogenesis. It acts as a natural agonist of RAR nuclear receptors (IC50s of 14 nM for RARα/β/γ) and binds to PPARβ/δ with a Kd of 17 nM. This compound inhibits transcription factor Nrf2 through activation of retinoic acid receptor alpha.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS