Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retinoic acid ethyl ester-d5 | 78995-96-7 | 333.52 g·mol⁻¹ | C22H27D5O2 | 5 MG
SDP

Catalog No. 5000221485
Encompass_Preferred

Retinoic acid ethyl ester-d5 is a deuterium-labeled analogue of retinoic acid ethyl ester supplied for use as an internal standard and tracer in analytical chemistry. It provides a mass-shifted form of the parent compound for accurate quantification and metabolic studies, and is provided in a small package with recommended low-temperature storage to maintain stability.

  • Deuterium-labeled for stable isotope tracing and internal standard use.
  • Molecular formula C22H27D5O2 and molecular weight 333.52 g·mol⁻¹.
  • Intended for LC-MS/MS and other mass spectrometry quantification workflows.
  • Supplied as a 5 mg package suitable for analytical applications.
  • Storage recommendations: -20°C, protect from light, store under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-21485 Supplier Medchemexpress LLC Supplier No. HY141778S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Retinoic acid ethyl ester-d5 is a deuterium-labeled analogue of retinoic acid ethyl ester supplied for use as an internal standard and tracer in analytical chemistry. It provides a mass-shifted form of the parent compound for accurate quantification and metabolic studies, and is provided in a small package with recommended low-temperature storage to maintain stability.

  • Deuterium-labeled for stable isotope tracing and internal standard use.
  • Molecular formula C22H27D5O2 and molecular weight 333.52 g·mol⁻¹.
  • Intended for LC-MS/MS and other mass spectrometry quantification workflows.
  • Supplied as a 5 mg package suitable for analytical applications.
  • Storage recommendations: -20°C, protect from light, store under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.