Learn More
Medchemexpress LLC Retinoic acid ethyl ester-d5 | 78995-96-7 | 99.2% | C22H27D5O2 | 10MG
SDP
Catalog No. 5000261463
Encompass_Preferred
Retinoic acid ethyl ester-d5 is the deuterium-labeled isotopologue of retinoic acid ethyl ester supplied for use as an internal standard in analytical chemistry. It is intended for isotope-dilution LC-MS/MS and related workflows to enable accurate quantitation and method validation.
  • Deuterium-labeled internal standard for isotope-dilution mass spectrometry.
  • High isotopic enrichment to improve quantitation accuracy.
  • High purity suitable for analytical workflows.
  • Supplied as an oil to facilitate solution preparation.
  • Stable under recommended storage conditions for consistent performance.
Catalog No. 50-002-61463 Supplier Medchemexpress LLC Supplier No. HY141778S10MG
Retinoic acid ethyl ester-d5 is the deuterium-labeled isotopologue of retinoic acid ethyl ester supplied for use as an internal standard in analytical chemistry. It is intended for isotope-dilution LC-MS/MS and related workflows to enable accurate quantitation and method validation.
  • Deuterium-labeled internal standard for isotope-dilution mass spectrometry.
  • High isotopic enrichment to improve quantitation accuracy.
  • High purity suitable for analytical workflows.
  • Supplied as an oil to facilitate solution preparation.
  • Stable under recommended storage conditions for consistent performance.