Learn More
Medchemexpress LLC Retinoic acid ethyl ester-d5 | 78995-96-7 | C22H27D5O2 | 1 MG
SDP
Catalog No. 5000356533
Encompass_Preferred
Retinoic acid ethyl ester-d5 is the deuterium labeled Retinoic acid ethyl ester. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled
  • Used as tracers for quantitation during drug development
  • Affects pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-56533 Supplier Medchemexpress LLC Supplier No. HY141778S1MG
Retinoic acid ethyl ester-d5 is the deuterium labeled Retinoic acid ethyl ester. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled
  • Used as tracers for quantitation during drug development
  • Affects pharmacokinetic and metabolic profiles of drugs