Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retinoic Acid Receptor alpha Antibody (YA096) | Protein A affinity purified | Predicted band size: 51 kDa; Observed band size: 75 kDa | 10 UL
SDP

Catalog No. 5000397499
Encompass_Preferred

Retinoic Acid Receptor alpha Antibody (YA096) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Retinoic Acid Receptor alpha. This antibody is suitable for research applications such as Western Blot and Flow Cytometry, offering high specificity and sensitivity for endogenous targets.

  • Rabbit-derived, monoclonal
  • Human species reactivity
  • Predicted molecular weight: 51 kDa; Observed: 75 kDa
  • Protein A affinity purified
  • Non-conjugated IgG isotype
  • Formulated in 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, with 0.05% Sodium Azide as preservative
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Applications: WB (1:50-1:2000), FC (1:50-1:100)

Catalog No. 50-003-97499 Supplier Medchemexpress LLC Supplier No. HYP8030810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Retinoic Acid Receptor alpha Antibody (YA096) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed to target Retinoic Acid Receptor alpha. This antibody is suitable for research applications such as Western Blot and Flow Cytometry, offering high specificity and sensitivity for endogenous targets.

  • Rabbit-derived, monoclonal
  • Human species reactivity
  • Predicted molecular weight: 51 kDa; Observed: 75 kDa
  • Protein A affinity purified
  • Non-conjugated IgG isotype
  • Formulated in 1*TBS (pH7.4), 0.05% BSA, 40% Glycerol, with 0.05% Sodium Azide as preservative
  • Store at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Applications: WB (1:50-1:2000), FC (1:50-1:100)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.