Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retinoic Acid Receptor beta Antibody (YA1574) | 10 uL
SDP

Catalog No. 5000441230
Encompass_Preferred

Retinoic Acid Receptor beta Antibody (YA1574) is a rabbit-host antibody reactive to human samples. It has an observed molecular weight of 55 kDa and is supplied as a liquid solution.

  • Host: Rabbit
  • Reactivity: Human
  • Clonality: YA1574
  • SwissProt ID: P10826
  • Gene ID: 5915
  • Appearance: Liquid
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, IHC-P, ICC/IF
  • Dilution ratio: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:50-1:200
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles
  • Shipping: Shipping with blue ice
  • Caution: Product has not been fully validated for medical applications. For research use only

Catalog No. 50-004-41230 Supplier Medchemexpress LLC Supplier No. HYP8182910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Retinoic Acid Receptor beta Antibody (YA1574) is a rabbit-host antibody reactive to human samples. It has an observed molecular weight of 55 kDa and is supplied as a liquid solution.

  • Host: Rabbit
  • Reactivity: Human
  • Clonality: YA1574
  • SwissProt ID: P10826
  • Gene ID: 5915
  • Appearance: Liquid
  • Concentration: 1.0 mg/mL
  • Conjugation: Non-conjugated
  • Application: WB, IHC-P, ICC/IF
  • Dilution ratio: WB: 1:500-1:1000; IHC-P: 1:50-1:100; ICC/IF: 1:50-1:200
  • Formulation: Supplied in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles
  • Shipping: Shipping with blue ice
  • Caution: Product has not been fully validated for medical applications. For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.