Learn More
Medchemexpress LLC Retinoic acid (Standard) | 302-79-4 | 99.7% | 300.44 | 100 MG
SDP
Catalog No. 5000362574
Encompass_Preferred
Retinoic acid (Standard) is the analytical standard of Retinoic acid, intended for research and analytical applications. It is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis.
  • Natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ
  • Binds to PPARβ/δ with a Kd of 17 nM
  • Acts as an inhibitor of transcription factor Nrf2 through activation of retinoic acid receptor alpha
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: solid, light yellow to yellow in color
  • Initial source: endogenous metabolite
Catalog No. 50-003-62574 Supplier Medchemexpress LLC Supplier No. HY14649R100MG
Add to Cart
Add to Cart
Retinoic acid (Standard) is the analytical standard of Retinoic acid, intended for research and analytical applications. It is a metabolite of vitamin A that plays important roles in cell growth, differentiation, and organogenesis.
  • Natural agonist of RAR nuclear receptors, with IC50s of 14 nM for RARα/β/γ
  • Binds to PPARβ/δ with a Kd of 17 nM
  • Acts as an inhibitor of transcription factor Nrf2 through activation of retinoic acid receptor alpha
  • Used as a reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Appearance: solid, light yellow to yellow in color
  • Initial source: endogenous metabolite