Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retinol, acetate, 9-cis- | 29584-22-3 | 85.0% | 328.49 | 1 MG
SDP

Catalog No. 5000393605
Encompass_Preferred

9-cis-Retinol acetate is a retinal derivative developed for research applications. This compound is presented as a solid, typically white to off-white in appearance, with a molecular weight of 328.49. Its purity is 85.0%, making it suitable for various scientific investigations.

  • Retinal derivative.
  • Intended for research use only.
  • Molecular formula C22H32O2.
  • Appearance is solid, white to off-white.
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-93605 Supplier Medchemexpress LLC Supplier No. HY16760S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-cis-Retinol acetate is a retinal derivative developed for research applications. This compound is presented as a solid, typically white to off-white in appearance, with a molecular weight of 328.49. Its purity is 85.0%, making it suitable for various scientific investigations.

  • Retinal derivative.
  • Intended for research use only.
  • Molecular formula C22H32O2.
  • Appearance is solid, white to off-white.
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.