Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Retinol, acetate, 9-cis- | 29584-22-3 | 85.0% | 328.49 | 1 MG
SDP

Catalog No. 5000365174
Encompass_Preferred

9-cis-Retinol acetate (also known as 9-cis-Vitamin A acetate or Zuretinol acetate) is a retinal derivative. This white to off-white solid has a molecular weight of 328.49 and a chemical formula of C22H32O2. Intended for research use only, it is suggested to modulate the visual cycle in pharmacotherapy for retinal disease.

  • Synonyms: 9-cis-Vitamin A acetate, Zuretinol acetate
  • Description: Retinal derivative
  • Appearance: White to off-white solid
  • Application: Visual cycle modulator in retinal disease pharmacotherapy
  • Usage: For research use only
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature (continental US)

Catalog No. 50-003-65174 Supplier Medchemexpress LLC Supplier No. 1MGHY16760
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9-cis-Retinol acetate (also known as 9-cis-Vitamin A acetate or Zuretinol acetate) is a retinal derivative. This white to off-white solid has a molecular weight of 328.49 and a chemical formula of C22H32O2. Intended for research use only, it is suggested to modulate the visual cycle in pharmacotherapy for retinal disease.

  • Synonyms: 9-cis-Vitamin A acetate, Zuretinol acetate
  • Description: Retinal derivative
  • Appearance: White to off-white solid
  • Application: Visual cycle modulator in retinal disease pharmacotherapy
  • Usage: For research use only
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping: Room temperature (continental US)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.