Learn More
Crystal Chem Inc Retinol Binding Protein (RBP4) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80733
Encompass
The Retinol Binding Protein ELISA is used to quantify levels of human retinol binding protein (RBP4) in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 1.17 ng/mL. Dynamic range of 1.56 - 100 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7851