Learn More
Athens Research And Technology Inc RETINOL BINDING PROTEIN HU 100
Supplier:  Athens Research And Technology Inc 1616180216100UG
Encompass_Preferred
Retinol Binding Protein (RBP4), Human Plasma: 100 UG; Lyophilized from 50 mM Tris, 500 mM NaCl, pH 7.4; MW 21,000; EC= 1.86; Purity: > =98% by SDS-PAGE. Native; Purified from human plasma in the Athens Research & Tech lab. Molecular Biology Reagent
Catalog No. 50-489-904