Learn More
Arbor Assays Retinol Binding Protein (RBP) Multi-format ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K062H1
Encompass_Preferred
The DetectX Retinol Binding Protein (RBP) Multi-Format ELISA Kits are designed to allow for the measurement of both high and low RBP levels in a variety of samples including dried blood spots, serum, EDTA and heparin plasma and urine with the same kit
Catalog No. 50-227-8491