Learn More
Medchemexpress LLC HY-N0679 100mg Medchemexpress, Retinyl acetate CAS:127-47-9 Purity:>98%
SDP
Catalog No. 501933671
Encompass_Preferred
Medchemexpress, HY-N0679 100mg Retinyl acetate CAS:127-47-9 Retinyl acetate is a natural form of vitamin A and has potential antineoplastic and chemo preventive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3671 Supplier Medchemexpress LLC Supplier No. HYN0679100MG
Medchemexpress, HY-N0679 100mg Retinyl acetate CAS:127-47-9 Retinyl acetate is a natural form of vitamin A and has potential antineoplastic and chemo preventive activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.