Learn More
Medchemexpress LLC Retinyl acetate | 127-47-9 | 328.49 | 1 ML
SDP
Catalog No. 5000388963
Encompass_Preferred
Retinyl acetate is a synthetic acetate ester form derived from retinol that possesses potential antineoplastic and chemopreventive activities. It is for research use only and not sold to patients.
  • Purity of 99.30%
  • Off-white to light yellow solid appearance
  • Classified as terpenoids, diterpenoids
  • Initial source is endogenous metabolite
  • Shipped at room temperature in continental US
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Inhibits proliferation of tumor cells at 10 μM in vitro
  • Inhibits DMBA induced mammary carcinogenesis in Sprague-Dawley rats at 250 mg/kg in diet in vivo
Catalog No. 50-003-88963 Supplier Medchemexpress LLC Supplier No. HYN067910MM/1ML
Retinyl acetate is a synthetic acetate ester form derived from retinol that possesses potential antineoplastic and chemopreventive activities. It is for research use only and not sold to patients.
  • Purity of 99.30%
  • Off-white to light yellow solid appearance
  • Classified as terpenoids, diterpenoids
  • Initial source is endogenous metabolite
  • Shipped at room temperature in continental US
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Inhibits proliferation of tumor cells at 10 μM in vitro
  • Inhibits DMBA induced mammary carcinogenesis in Sprague-Dawley rats at 250 mg/kg in diet in vivo