Learn More
Medchemexpress LLC Retinyl acetate-d4 | 118139-40-5 | 332.51 | 1 MG
SDP
Catalog No. 5000389037
Encompass_Preferred
Retinyl acetate-d4 is the deuterium-labeled Retinyl acetate. Retinyl acetate is a synthetic acetate ester form derived from retinol and has potential antineoplastic and chemopreventive activities. This compound has potential antineoplastic and chemopreventive activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89037 Supplier Medchemexpress LLC Supplier No. HYN0679S41MG
Retinyl acetate-d4 is the deuterium-labeled Retinyl acetate. Retinyl acetate is a synthetic acetate ester form derived from retinol and has potential antineoplastic and chemopreventive activities. This compound has potential antineoplastic and chemopreventive activities.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.