Learn More
American Radiolabeled Chemicals Inc RETINYL PALM-9 10-3H 250UC
SDP
Catalog No. 5090105075
Encompass_Preferred
Retinyl palmitate-palmitoyl-9,10-3H 250uCi in toluene, 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 524.9, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-05075 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1420250UC
Add to Cart
Add to Cart
Request a Quote
Retinyl palmitate-palmitoyl-9,10-3H 250uCi in toluene, 1 mCi/ml, S.A. 50-60 Ci/mmol, M.W. 524.9, Shipped in Dry Ice, Exclusive