Learn More
Medchemexpress LLC HY-B1384 100mg Medchemexpress, Retinyl (palmitate) CAS:79-81-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1384100MG
Encompass_Preferred
Medchemexpress, HY-B1384 100mg Retinyl (palmitate) CAS:79-81-2 Retinyl palmitate is an ester of Retinol and is the major form of vitamin A found in the epidermis. Retinyl palmitate has been widely used in pharmaceutical and cosmetic formulations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0149