Learn More
Sigma Aldrich Fine Chemicals Biosciences Retinyl propionate 98.0 su100G
Catalog No. 502899479
Encompass
Retinal retinol and retinoic acid are the aldehyde alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinoids also exist as retinyl esters such as retinyl propionate retinyl acetate and retinyl palmitate.
Catalog No. 50-289-9479 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 87809100G
Retinal retinol and retinoic acid are the aldehyde alcohol and acid forms of vitamin A. The retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinoids also exist as retinyl esters such as retinyl propionate retinyl acetate and retinyl palmitate.