Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinyl Propionate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015306 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5306 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5306 Supplier MilliporeSigma Supelco Supplier No. PHR16681G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Retinyl Propionate belongs to the group of retinyl esters and is found to be active in human skin. It shows cosmetic related efficacy and hence helps in the improvement of photodamaged facial skin.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7069-42-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Liquid
Linear Formula C23H34O2
MDL Number MFCD00129785
Synonym Retinyl propionate; Vitamin A propionate
Type Certified Reference Material
Quantity 1 g
Formula Weight 342.51
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.