Learn More
Apexbio Technology LLC Retinyl (Vitamin A) Palmitate 79-81-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094296
Encompass_Preferred
Retinyl Vitamin A Palmitate is a stabilized ester derivative of retinol (Vitamin A) and palmitic acid It is designed to deliver bioavailable Vitamin A upon enzymatic hydrolysis releasing free retinol that functions as an active intermediate in visual function cellular differentiation gene expression regulation and immune responses via retinoid receptors Retinyl Vitamin A Palmitate exerts its biological activity primarily through enzymatic hydrolysis generating retinol that participates in retinoid-mediated signaling pathways Based on these pharmacological properties Retinyl Vitamin A Palmitate holds research potential in studies of retinoid signaling vitamin deficiency models dermatological processes and nutritional supplementation evaluations
Catalog No. 50-000-94296 Supplier Apexbio Technology LLC Supplier No. B18695.1
Retinyl Vitamin A Palmitate is a stabilized ester derivative of retinol (Vitamin A) and palmitic acid It is designed to deliver bioavailable Vitamin A upon enzymatic hydrolysis releasing free retinol that functions as an active intermediate in visual function cellular differentiation gene expression regulation and immune responses via retinoid receptors Retinyl Vitamin A Palmitate exerts its biological activity primarily through enzymatic hydrolysis generating retinol that participates in retinoid-mediated signaling pathways Based on these pharmacological properties Retinyl Vitamin A Palmitate holds research potential in studies of retinoid signaling vitamin deficiency models dermatological processes and nutritional supplementation evaluations