Learn More
Apexbio Technology LLC Retinyl (Vitamin A) Palmitate 79-81-2 50mg
SDP
Catalog No. 5000089856
Encompass_Preferred
Retinyl Vitamin A Palmitate is a stabilized ester derivative of retinol (Vitamin A) and palmitic acid It is designed to deliver bioavailable Vitamin A upon enzymatic hydrolysis releasing free retinol that functions as an active intermediate in visual function cellular differentiation gene expression regulation and immune responses via retinoid receptors Retinyl Vitamin A Palmitate exerts its biological activity primarily through enzymatic hydrolysis generating retinol that participates in retinoid-mediated signaling pathways Based on these pharmacological properties Retinyl Vitamin A Palmitate holds research potential in studies of retinoid signaling vitamin deficiency models dermatological processes and nutritional supplementation evaluations
Catalog No. 50-000-89856 Supplier Apexbio Technology LLC Supplier No. B186950
Retinyl Vitamin A Palmitate is a stabilized ester derivative of retinol (Vitamin A) and palmitic acid It is designed to deliver bioavailable Vitamin A upon enzymatic hydrolysis releasing free retinol that functions as an active intermediate in visual function cellular differentiation gene expression regulation and immune responses via retinoid receptors Retinyl Vitamin A Palmitate exerts its biological activity primarily through enzymatic hydrolysis generating retinol that participates in retinoid-mediated signaling pathways Based on these pharmacological properties Retinyl Vitamin A Palmitate holds research potential in studies of retinoid signaling vitamin deficiency models dermatological processes and nutritional supplementation evaluations