Learn More
Aviva Systems Biology Corp RETNLB Antibody - N-terminal region (ARP60355_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60355_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RETNLB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RETNLB. Alternate names: FIZZ1, FIZZ2, HXCP2, RELM-beta, RELMb, RELMbeta, XCP2ApplicationWB. Species reactivity: Human
Catalog No. 50-287-018