Learn More
US Plastic Corporation Two-Prong Vinyl-Coated Retort Clamp with Boss Head
SDP
Catalog No. NC3308061
Encompass
Ideal for use with beakers, Erlenmeyer flasks, test tubes, boiling (Florence) flasks, and burettes, these retort clamps include a high-strength alloy boss head to secure the clamp to a frame or other apparatus. The clamps, constructed of solid die-cast zinc with a blue powder coating, have a metal adjustment screw that allows for precise pressure when gripping glass objects to avoid potential breakage. Each clamp is non-ferrous, rust-free, and long-lasting in any laboratory application and contains a swivel between the clamp and the boss head. The boss head will accept a rod up to 18mm in diameter, and, if needed, the boss head and clamp can be separated with a 5/32 Allen wrench. The vinyl-coated clamps (23458, 23460, 23461) are excellent for higher temperature applications, while it is not recommended that the cork-lined clamps (23459, 23466, 23467) be placed in direct heat.
Catalog No. NC3308061 Supplier US Plastic Corporation Supplier No. 23461
Ideal for use with beakers, Erlenmeyer flasks, test tubes, boiling (Florence) flasks, and burettes, these retort clamps include a high-strength alloy boss head to secure the clamp to a frame or other apparatus. The clamps, constructed of solid die-cast zinc with a blue powder coating, have a metal adjustment screw that allows for precise pressure when gripping glass objects to avoid potential breakage. Each clamp is non-ferrous, rust-free, and long-lasting in any laboratory application and contains a swivel between the clamp and the boss head. The boss head will accept a rod up to 18mm in diameter, and, if needed, the boss head and clamp can be separated with a 5/32 Allen wrench. The vinyl-coated clamps (23458, 23460, 23461) are excellent for higher temperature applications, while it is not recommended that the cork-lined clamps (23459, 23466, 23467) be placed in direct heat.