Description
- Includes a protective tube.
- FS019 includes a pocket clip.
- Use with any magnetic print powder
- Ideal for dusting on Non-ferrous surfaces such as paper, wood, leather, and plastic.
- Includes a protective storage tube for safe handling.
Specifications
Specifications
| Product Type | Magnetic Wand |
| Includes | Protective Storage Tube |
| Format | Regular |