Learn More
Mopec Retractable Tip Evidence Scriber
SDP
Catalog No. 22039064 Shop All Mopec Products
Change view
Click to view available options
Tip Material:
Carbide
Diamond
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Material
22-039-064 Diamond
22-039-062 Carbide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-039-064 Supplier Mopec Supplier No. FS839
Add to Cart
Add to Cart
Built for long-term use, replacement tips are available when needed.
  • formance
    • Used for marking hard surfaces such as metal, glass, and plastic
  • Construction
    • Retractable tip
    • Carbide or diamond tips
  • Materials
    • Carbide
    • Diamond
Specifications
Product Type Scribe Pen
Tip Style Retractable
Color Silver
Autoclavable Autoclavable
Sterility Non-sterile
Tip Material Diamond
Disposable Reusable
Latex Free Yes