Learn More
Swann Morton Retracting Blade Safety Scalpel
SDP
Catalog No. 01001376
Change view
Click to view available options
Size:
# 11P
Catalog No. Size
01-001-376 # 11P
1 options
Catalog No. 01-001-376 Supplier Swann Morton Supplier No. 05SM11P
Add to Cart
Add to Cart
Sterile retracting blade safety scalpel with a stainless steel blade for safe and controlled cutting.
  • Supports safe passing procedures and helps follow safety protocols
  • Stainless Steel
  • Designed to improve safe handling during use
  • Color coded for quick and easy blade shape recognition
Specifications
Product Type Scalpel
Size # 11P
Color Green