Learn More
Swann Morton Retracting Guard Safety Scalpel
SDP
Catalog No. 01001119
Change view
Click to view available options
Size:
# 10
# 11
# 11P
# 15
# 20
# 21
# 22
# 23
# 24
Sterility:
Non-sterile
Sterile
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility
01-001-119 # 10 Non-sterile
01-001-214 # 10 Sterile
01-001-124 # 11 Non-sterile
01-001-220 # 11 Sterile
01-001-212 # 11P Non-sterile
01-001-320 # 11P Sterile
01-001-127 # 15 Non-sterile
01-001-226 # 15 Sterile
01-001-130 # 20 Non-sterile
01-001-248 # 20 Sterile
01-001-137 # 21 Non-sterile
01-001-258 # 21 Sterile
01-001-202 # 22 Non-sterile
01-001-280 # 22 Sterile
01-001-204 # 23 Non-sterile
01-001-308 # 23 Sterile
01-001-210 # 24 Non-sterile
01-001-316 # 24 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 01-001-119 Supplier Swann Morton Supplier No. SM3501
Add to Cart
Add to Cart
This item is not returnable. View return policy
Retracting guard safety scalpel available in a variety of sizes.
  • Stainless Steel
  • Designed for tissue separation, puncturing or cutting procedures
  • Easy one-handed activation
  • OSHA compliant
  • Covered blades reduces risk of accidental contact
Specifications
Product Type Safety Scalpel
Size # 10
Disposable Single-use
Sterility Non-sterile
Autoclavable Autoclavable
Graduated Yes