Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Swann Morton Retracting Guard Safety Scalpel
SDP

Catalog No. 01001320
Change view
Click to view available options
Size:
# 10
# 11
# 11P
# 15
# 20
# 21
# 22
# 23
# 24
Sterility:
Non-sterile
Sterile
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Sterility
01-001-320 # 11P Sterile
01-001-119 # 10 Non-sterile
01-001-214 # 10 Sterile
01-001-124 # 11 Non-sterile
01-001-220 # 11 Sterile
01-001-212 # 11P Non-sterile
01-001-127 # 15 Non-sterile
01-001-226 # 15 Sterile
01-001-130 # 20 Non-sterile
01-001-248 # 20 Sterile
01-001-137 # 21 Non-sterile
01-001-258 # 21 Sterile
01-001-202 # 22 Non-sterile
01-001-280 # 22 Sterile
01-001-204 # 23 Non-sterile
01-001-308 # 23 Sterile
01-001-210 # 24 Non-sterile
01-001-316 # 24 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 01-001-320 Supplier Swann Morton Supplier No. SM3691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Retracting guard safety scalpel available in a variety of sizes.

  • Stainless Steel
  • Designed for tissue separation, puncturing or cutting procedures
  • Easy one-handed activation
  • OSHA compliant
  • Covered blades reduces risk of accidental contact

Specifications

Product Type Safety Scalpel
CAS 67-56-1
Size # 11P
Disposable Single-use
Sterility Sterile
Autoclavable Autoclavable
Graduated Yes
Linear Formula 13C3C15H22O2
MDL Number MFCD15144880
Concentration 10 μg/mL in methanol
Formula Weight 273.34
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.