Learn More
Fine Science Tools RETRACTOR 16MM DEEP 7CM LONG
SDP
Catalog No. NC9196416
Encompass
Product: Alm Retractor - 4x4 Blunt/7cm/4cm Maximum Spread/16mmTeeth Length/ Teeth: 4x4 Blunt/ Teeth Width: 7mm/ Teeth Depth: 16mm/ Alloy: Stainless Steel/ Length: 7cm/ Unit: Each
Catalog No. NC9196416 Supplier Fine Science Tools Supplier No. 1700907
Product: Alm Retractor - 4x4 Blunt/7cm/4cm Maximum Spread/16mmTeeth Length/ Teeth: 4x4 Blunt/ Teeth Width: 7mm/ Teeth Depth: 16mm/ Alloy: Stainless Steel/ Length: 7cm/ Unit: Each