Learn More
Fine Science Tools Alm Retractors, 4 x 4 Blunt, 7 cm L, each
SDP
Catalog No. NC9131905
Encompass
Product: Alm Retractor - 4x4 Blunt/7cm/4cm Maximum Spread/5.5mm Teeth Length/ Teeth: 4 Pronged Blunt/ Teeth Length: / Teeth Width: 7mm/ Teeth Depth: 3.5mm/ Alloy: Stainless Steel/ Length: 7cm/ Unit: Each
Catalog No. NC9131905 Supplier Fine Science Tools Supplier No. 1700807
Product: Alm Retractor - 4x4 Blunt/7cm/4cm Maximum Spread/5.5mm Teeth Length/ Teeth: 4 Pronged Blunt/ Teeth Length: / Teeth Width: 7mm/ Teeth Depth: 3.5mm/ Alloy: Stainless Steel/ Length: 7cm/ Unit: Each