Learn More
World Precision Instrument ALM Self-Retaining Retractor, 7cm, 4x4 prongs
SDP
Catalog No. 50822304
Encompass
The ALM (Andrews-Lucia-McPherson) self-retaining retractor is a surgical instrument designed to hold the eyelids open during ophthalmic surgery, providing surgeons with a clear view of the eye. The two blades have a curved end that fits into the fornices of the eye. The adjustable blades are held in place by a spring-loaded mechanism. The ALM retractor was designed in the 1960s by three ophthalmologist. It is crafter of surgical grade stainless steel or premium German steel, and they may be autoclaved or sterilized using a chemical sterilant. Clean your surgical instruments promptly after every use to ensure a long life for your surgical tools.
Catalog No. 50-822-304 Supplier World Precision Instrument Supplier No. 14240
The ALM (Andrews-Lucia-McPherson) self-retaining retractor is a surgical instrument designed to hold the eyelids open during ophthalmic surgery, providing surgeons with a clear view of the eye. The two blades have a curved end that fits into the fornices of the eye. The adjustable blades are held in place by a spring-loaded mechanism. The ALM retractor was designed in the 1960s by three ophthalmologist. It is crafter of surgical grade stainless steel or premium German steel, and they may be autoclaved or sterilized using a chemical sterilant. Clean your surgical instruments promptly after every use to ensure a long life for your surgical tools.