Learn More
Fine Science Tools Magnetic Fixator Retraction System, 2.5 cm x 2.5 mm
SDP
Catalog No. NC9222292
Encompass
Product: Magnetic Fixator Retraction System - Blunt Retractors/2.5mm Wide/ Item: Kit for Animals 200g to 5kg/ Maximum Spread: Adjustable
Catalog No. NC9222292 Supplier Fine Science Tools Supplier No. 1820010
Product: Magnetic Fixator Retraction System - Blunt Retractors/2.5mm Wide/ Item: Kit for Animals 200g to 5kg/ Maximum Spread: Adjustable