Learn More
Med Vet International Miltex Ribbon Retractor, 3/4in. X 13in.
Catalog No. 502832161
Encompass_Preferred
The Miltex 11-130 Ribbon Retractor is a malleable retractor that measure 3/4in. Wide and 13in. in length. It is made of German grade stainless steel and can be used to retract deep wounds and may be bent to various shapes.
Catalog No. 50-283-2161 Supplier Med Vet International Supplier No. MIL11130
The Miltex 11-130 Ribbon Retractor is a malleable retractor that measure 3/4in. Wide and 13in. in length. It is made of German grade stainless steel and can be used to retract deep wounds and may be bent to various shapes.