Learn More
Med Vet International Miltex Vantage Senn Retractor, 6.5in., Sharp Prongs, Double Ended
Catalog No. 502830413
Encompass_Preferred
The Miltex Vantage V911-74 Senn Retractor is made of mid-grade stainless steel and has a length of 6-3/8in. This double ended Senn Retractor is delicate retractor that is commonly used in hand surgery, vascular surgery, plastic surgery and other procedures involving the skin and soft tissue.
Catalog No. 50-283-0413 Supplier Med Vet International Supplier No. MILV91174
The Miltex Vantage V911-74 Senn Retractor is made of mid-grade stainless steel and has a length of 6-3/8in. This double ended Senn Retractor is delicate retractor that is commonly used in hand surgery, vascular surgery, plastic surgery and other procedures involving the skin and soft tissue.