Learn More
Med Vet International Miltex Senn Retractor, 6.25in., Blunt
Catalog No. 502830100
Encompass_Preferred
The Miltex 11-76 Senn Retractor is made of German grade stainless steel and has a length of 6-3/8in. This double ended Senn Retractor has 3 Blunt 6.5 X 19 mm Prongs and 9.5 mm Blade. It is a delicate retractor that is commonly used in hand surgery, vascular surgery, plastic surgery and other procedures involving the skin and soft tissue.
Catalog No. 50-283-0100 Supplier Med Vet International Supplier No. MIL1176
The Miltex 11-76 Senn Retractor is made of German grade stainless steel and has a length of 6-3/8in. This double ended Senn Retractor has 3 Blunt 6.5 X 19 mm Prongs and 9.5 mm Blade. It is a delicate retractor that is commonly used in hand surgery, vascular surgery, plastic surgery and other procedures involving the skin and soft tissue.