Learn More
Med Vet International Miltex U.S. Army Retractor, 8.5in, 2/Set
Catalog No. 502831189
Encompass_Preferred
The Miltex 11-122 Retractor is made of German grade stainless steel. It is a double ended retractor with a flat handle and a length of 8.5in.
Catalog No. 50-283-1189 Supplier Med Vet International Supplier No. MIL11122
The Miltex 11-122 Retractor is made of German grade stainless steel. It is a double ended retractor with a flat handle and a length of 8.5in.