Learn More
World Precision Instrument Wire Retractor, 5cm, 15x10mm Blades
SDP
Catalog No. 50822479
Encompass
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our small wire speculum is made of surgical grade stainless steel or our premium German steel, and it comes in two sizes, with a spread of 13 and 25 mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.
Catalog No. 50-822-479 Supplier World Precision Instrument Supplier No. 500368
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our small wire speculum is made of surgical grade stainless steel or our premium German steel, and it comes in two sizes, with a spread of 13 and 25 mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.