Learn More
Med Vet International U.S. Army Retractor, 8.25in, Double Ended, German Grade, Von Klaus
Catalog No. 502092659
Encompass_Preferred
8 1/4' US Army Retractor is a German grade surgical instrument and come in a set of 2. they are a double ended retractor with blades at 90 degrees facing the same direction. US Army Retractors are commonly used to expose surface layers of the skin. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2659 Supplier Med Vet International Supplier No. VK1221821
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
8 1/4' US Army Retractor is a German grade surgical instrument and come in a set of 2. they are a double ended retractor with blades at 90 degrees facing the same direction. US Army Retractors are commonly used to expose surface layers of the skin. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.