Learn More
Med Vet International Petite Balfour Retractor 4" spread German
Catalog No. 502092667
Encompass_Preferred
Fenestrated side blades and solid center blade 1" deep, 4" spreadThe Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2667 Supplier Med Vet International Supplier No. VK025170A
Fenestrated side blades and solid center blade 1" deep, 4" spreadThe Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.