Learn More
Med Vet International Balfour Abdominal Retractor, 7.375in, Fixed Fenestrated Side Blades, Von Klaus German Surgical Steel
Catalog No. 502092674
Encompass_Preferred
Features a fixed side blade, fixed fenestrated side blade, 2.5' deep center blade that is 3'x2', and has a 7.375 spread. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2674 Supplier Med Vet International Supplier No. VK1247010
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Features a fixed side blade, fixed fenestrated side blade, 2.5' deep center blade that is 3'x2', and has a 7.375 spread. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.