Learn More
Med Vet International Miltex Allison Lung Retractor, 12.75in, Standard Size: Blade 2in Wide, German, 25-302
Catalog No. 502829542
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Allison Lung Retractor. Retractors help surgeons and operating room professionals hold an incision or wound open during surgical procedures. They aid in holding back underlying organs or tissues, allowing doctors/nurses better visibility and access to the exposed area. This is made from premium-grade German stainless steel. Specifications: 12.75in, Standard Size: Blade 2in wide
Catalog No. 50-282-9542 Supplier Med Vet International Supplier No. MIL25302
 Restricted Item
Add to Cart
Add to Cart
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Allison Lung Retractor. Retractors help surgeons and operating room professionals hold an incision or wound open during surgical procedures. They aid in holding back underlying organs or tissues, allowing doctors/nurses better visibility and access to the exposed area. This is made from premium-grade German stainless steel. Specifications: 12.75in, Standard Size: Blade 2in wide