Learn More
Med Vet International Miltex Rigid Neck Rake Retractor, 6.5in, 2 Sharp Prongs, German, 11-52
Supplier:  Med Vet International MIL1152
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Rigid Neck Rake Retractor. Retractors help surgeons and operating room professionals hold an incision or wound open during surgical procedures. They aid in holding back underlying organs or tissues, allowing doctors/nurses better visibility and access to the exposed area. This is made from premium-grade German stainless steel. Specifications: 6.5in, 2 Sharp Prongs
Catalog No. 50-283-2044