Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Retrieval System 50 ft 310 lb

Supplier:  Grainger 19TJ88

Encompass_Preferred

Retrieval System 50 ft 310 lb Yellow

Catalog No. 50-279-5726


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.