Learn More
Grainger Gemtor Retrieval Winch, Three-Way, Stainless Steel, 100 ft., 1 ea
Catalog No. 502795569
Encompass_Preferred
The Sentry RS3 series is a fully integrated three-function device designed for fall arrest, retrieval, and lowering applications. This system is engineered for use in OSHA-defined confined spaces, such as manholes, tanks, and vats, providing reliable performance in rescue operations.
  • Integrated fall arrest, retrieval, and lowering capabilities in a single unit
  • Two independent braking systems, including an energy-absorbing centrifugal brake that arrests falls within 24 in.
  • Friction brake that activates immediately to hold the load in place when raising or lowering stops
  • Designed with two handle positions for varying levels of retrieval force and speed
  • Constructed with a 100 ft. stainless steel cable
  • Meets OSHA and ANSI Z359.1 standards
  • Equipped with a stress indicator for safety monitoring
Catalog No. 50-279-5569 Supplier Grainger Supplier No. 19TJ82
Add to Cart
Add to Cart
Request a Quote
The Sentry RS3 series is a fully integrated three-function device designed for fall arrest, retrieval, and lowering applications. This system is engineered for use in OSHA-defined confined spaces, such as manholes, tanks, and vats, providing reliable performance in rescue operations.
  • Integrated fall arrest, retrieval, and lowering capabilities in a single unit
  • Two independent braking systems, including an energy-absorbing centrifugal brake that arrests falls within 24 in.
  • Friction brake that activates immediately to hold the load in place when raising or lowering stops
  • Designed with two handle positions for varying levels of retrieval force and speed
  • Constructed with a 100 ft. stainless steel cable
  • Meets OSHA and ANSI Z359.1 standards
  • Equipped with a stress indicator for safety monitoring