Learn More
Electron Microscopy Sciences Retriever Thermal Slide Processor, 220 V
SDP
Catalog No. 5031170
Encompass_Preferred
Retriever that solves the problem of staining formalin fixed tissues. Affordable solution to all known major problems with immunohistochemistry on paraffin sections. Ease of use combined with high reproducibility of the results will give you the best quality immunostaining.
Bench-top model for thermally processing slides of formalin-fixed, paraffin embedded tissues prior to immunostaining. Designed to ensure identical processing of all the samples during a processing cycle, as well as the identical processing of the samples in individual sessions. Preserves processed tissues.
Catalog No. 50-311-70 Supplier Electron Microscopy Sciences Supplier No. 6270020
Add to Cart
Add to Cart
Retriever that solves the problem of staining formalin fixed tissues. Affordable solution to all known major problems with immunohistochemistry on paraffin sections. Ease of use combined with high reproducibility of the results will give you the best quality immunostaining.
Bench-top model for thermally processing slides of formalin-fixed, paraffin embedded tissues prior to immunostaining. Designed to ensure identical processing of all the samples during a processing cycle, as well as the identical processing of the samples in individual sessions. Preserves processed tissues.