Learn More
Electron Microscopy Sciences Retriever Tissue Adhesive Slides
SDP
Catalog No. 501927054
Encompass_Preferred
Retriever Tissue Slides are high adhesive glass microscope slides for paraffin-embedded tissue section, including tissue arrays. The slides were designed and optimized for the EMS Antigen Retriever (62700 series) to preserve the section attachment and tissue morphology during heat-induced epitope recovery in Retriever in pH6 (Low), pH9 (High), or Universal Retriever buffer.
Slides can also be successfully used in any other epitope-recovery technology (autoclave, microwave, proteolitic) and show superior performance over positively charged or polylysine-coated slides, especially in high pH buffers and recovery buffers containing EDTA.
Catalog No. 50-192-7054 Supplier Electron Microscopy Sciences Supplier No. 7188050CS
Retriever Tissue Slides are high adhesive glass microscope slides for paraffin-embedded tissue section, including tissue arrays. The slides were designed and optimized for the EMS Antigen Retriever (62700 series) to preserve the section attachment and tissue morphology during heat-induced epitope recovery in Retriever in pH6 (Low), pH9 (High), or Universal Retriever buffer.
Slides can also be successfully used in any other epitope-recovery technology (autoclave, microwave, proteolitic) and show superior performance over positively charged or polylysine-coated slides, especially in high pH buffers and recovery buffers containing EDTA.