Learn More
Electron Microscopy Sciences Retrieving Tweezers, 12"L
SDP
Catalog No. 50996744
Encompass_Preferred
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.
Catalog No. 50-996-744 Supplier Electron Microscopy Sciences Supplier No. 6211212
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.