Learn More
Electron Microscopy Sciences Retrieving Tweezers 6"
SDP
Catalog No. 50996741
Encompass_Preferred
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.
Catalog No. 50-996-741 Supplier Electron Microscopy Sciences Supplier No. 6211206
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.