Learn More
Electron Microscopy Sciences Retrieving Tweezers, 8"L
SDP
Catalog No. 50996742
Encompass_Preferred
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.
Catalog No. 50-996-742 Supplier Electron Microscopy Sciences Supplier No. 6211208
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.